Learn More
Glassco Laboratory Equipments Erlenmeyer Glass Flask Heavy duty
Descrizione
- Heavy-duty Erlenmeyer glass flasks with narrow mouths
- Available in 50 mL, 100 mL, 250 mL, 500 mL, 1000 mL, and 2000 mL capacities
- Made from heat-resistant ASTM E-438 Type 1, Boro 3.3 glass
- Heavy-wall construction
- Complies with ISO 1773
- Printed with durable white enamel
- Reinforced thick rim helps reduce chipping and breakage
- Chemical resistant
Specifica
Specifica
| Capacità (sistema metrico) | 1000 mL |
| Materiale | Borosilicate Glass (3.3) |
| Product Type | Heavy-duty Erlenmeyer Flask |
| Autoclavabile | Autoclavable |
| Monouso | Reusable |
| Graduated | Yes |
| Mouth Style | Narrow |
| Sterilità | Non-sterile |
| Forma del fondo | Flat |
| Trattamento superficiale | None |