Learn More
Ethyl 3-isocyanatopropionate, 98%
Codice prodotto. p-7045152
Cambia vista
Click to view available options
Quantità:
1 g
5 g
Dimensione della confezione:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11322167 1 g 1g
11332167 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11322167 Fornitore Thermo Scientific Alfa Aesar N. del fornitore L09616.03
Richiesta di formato massivo o personalizzato
Please to purchase this item. Need a web account? Register with us today!
CAS: 5100-34-5 | C6H9NO3 | 143.14 g/mol
Ethyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethyl 3-isocyanatopropionate can be used to produce 3-[1-(2-dimethylamino-ethylcarbamoyl)-5,6-dimethyl-6H-pyrido[4,3-b]carbazol-9-yloxycarbonylamino]-propionic acid ethyl ester with 9-hydroxy-5,6-dimethyl-6H-pyrido[4,3-b]carbazole-1-carboxylic acid (2-dimethylamino-ethyl)-amide.
Solubility
Not miscible with water.
Notes
Store at 4°C. Moisture Sensitive. Avoid moisture, heat and direct sunlight. Incompatible materials are amines, alcohols and heat. Store away from moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemische Identifikatoren
CAS 5100-34-5
Formula molecolare C6H9NO3
Molecular Weight (g/mol) 143.14
Numero MDL MFCD00078461
InChI Key XBSGYVHOINMTIM-UHFFFAOYSA-N
Sinonimo ethyl 3-isocyanatopropionate, propanoic acid, 3-isocyanato-, ethyl ester, 3-isocyanatopropionic acid ethyl ester, ethyl n-oxomethylidene-beta-alaninate, ethyl n-oxomethylene-beta-alaninate, acmc-20akzk, ethyl-3-isocyanatopropionate, ethyl 3-isocyanato-propionate, ethyl 3-isocyanatopropion-ate, ethyl-3-isocyanato-propionate
PubChem CID 2733373
IUPAC Name ethyl 3-isocyanatopropanoate
SMILES CCOC(=O)CCN=C=O
Spezifikation
Materiale o nome chimico Ethyl 3-isocyanatopropionate
Densità 1.087
Punti di ebollizione 74°C to 75°C (4 mmHg)
Punto d'infiammabilità 92°C (197°F)
Odore Irritating
Indice di rifrazione 1.429
Quantità 1 g
Numero UN UN2206
Beilstein 1705618
Sensibilità Moisture sensitive
Informazioni di solubilità Not miscible with water.
Peso formulazione 143.14
Percent Purity 98%
Mehr anzeigen Weniger anzeigen
RUO – Research Use Only
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.