Learn More
Ethyl 4,4,4-trifluoro-2-methylacetoacetate, 96%
Product Code. 11378386
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11378386 5 g 5g
11368386 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11378386 Supplier Thermo Scientific Alfa Aesar Supplier No. L17335.06
 Restricted Item
Spedizione stimata: 30-11-2026
Add to basket
CAS: 344-00-3 | C7H9F3O3 | 198.141 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 344-00-3
Molecular Formula C7H9F3O3
Molecular Weight (g/mol) 198.141
MDL Number MFCD00190635
InChI Key YLRGPBKEZVHOAW-UHFFFAOYSA-N
Synonym ethyl 2-methyl-4,4,4-trifluoroacetoacetate, butanoic acid, 4,4,4-trifluoro-2-methyl-3-oxo-, ethyl ester, zlchem 125, acmc-20al0w, ethyl 2-methyltrifluoroacetoacetate, ethyl 2-methyl-4,4,4-trifluoro acetoacetate, ethyl alpha-methyl-4,4,4-trifluoroacetoacetate, ethyl4,4,4-trifluoro-2-methyl-3-oxobutanoate, ethyl 3-keto-2-methyl-4,4,4-trifluorobutanoate, ethyl 4,4,4-trifluoro-2-methyl-3-oxo-butanoate
PubChem CID 2737204
IUPAC Name ethyl 4,4,4-trifluoro-2-methyl-3-oxobutanoate
SMILES CCOC(=O)C(C)C(=O)C(F)(F)F
Tekniske data
Density 1.258
Boiling Point 144°C to 146°C
Flash Point 47°C (116°F)
Refractive Index 1.37
Quantity 5 g
UN Number UN3272
Beilstein 1783620
Formula Weight 198.14
Percent Purity 96%
Chemical Name or Material Ethyl 4,4,4-trifluoro-2-methylacetoacetate
RUO – Research Use Only
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.