missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4,6-dichloro-8-methylquinoline-3-carboxylate, 97%, Thermo Scientific™

Catalog Number p-7039949
Click to view available options
Quantità:
250 mg
1 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
338954-50-0
C13H11Cl2NO2
284.136
MFCD00173360
DHCZSIOJQCTCHL-UHFFFAOYSA-N
4,6-dichloro-8-methylquinoline-3-carboxylic ethyl ester, 4,6-dichloro-8-methyl-quinoline-3-carboxylic acid ethyl ester, pubchem6083, ethyl 4,6-dichloro-8-methyl-3-quinolinecarboxylate, ethyl 4,6-bis chloranyl-8-methyl-quinoline-3-carboxylate, 4,6-dichloro-8-methyl-3-quinolinecarboxylic acid ethyl ester, 4,6-dichloro-8-methylquinoline-3-carboxylic acid ethyl ester
730825
ethyl 4,6-dichloro-8-methylquinoline-3-carboxylate
CCOC(=O)C1=C(C2=CC(=CC(=C2N=C1)C)Cl)Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
338954-50-0
C13H11Cl2NO2
284.136
MFCD00173360
DHCZSIOJQCTCHL-UHFFFAOYSA-N
4,6-dichloro-8-methylquinoline-3-carboxylic ethyl ester, 4,6-dichloro-8-methyl-quinoline-3-carboxylic acid ethyl ester, pubchem6083, ethyl 4,6-dichloro-8-methyl-3-quinolinecarboxylate, ethyl 4,6-bis chloranyl-8-methyl-quinoline-3-carboxylate, 4,6-dichloro-8-methyl-3-quinolinecarboxylic acid ethyl ester, 4,6-dichloro-8-methylquinoline-3-carboxylic acid ethyl ester
730825
ethyl 4,6-dichloro-8-methylquinoline-3-carboxylate
CCOC(=O)C1=C(C2=CC(=CC(=C2N=C1)C)Cl)Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico Ethyl 4,6-dichloro-8-methylquinoline-3-carboxylate
Quantità 250 mg
Peso formulazione 284.14
Percent Purity 97%
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'