missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acrylate, 99%, stab. with ca 20ppm 4-methoxyphenol

Codice prodotto. 11432877
Cambia vista
Click to view available options
Quantità:
100 mL
500 mL
2500 mL
Dimensione della confezione:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
11432877 500 mL 500mL
11448940 100 mL 100mL
11442877 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11432877 Fornitore Thermo Scientific Alfa Aesar N. del fornitore A15493.AP
 Articolo soggetto a restrizioni
In stock
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

CAS: 140-88-5 | C5H8O2 | 100.12 g/mol

Ethyl acrylate is used in the syntheses of various pharmaceutical intermediates, textiles and non-woven fibers. It is widely used in the production of polymers including resins, plastics, rubber and denture material. It is actively involved in the preparation of dimethylaminoethylacrylate (DMAEA) which is used as a monomer to make flocculants for water treatment. It plays a vital role in the manufacture of water-based latex paints and adhesives, textile and paper coatings and leather finish resins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl acrylate is used in the syntheses of various pharmaceutical intermediates, textiles and non-woven fibers. It is widely used in the production of polymers including resins, plastics, rubber and denture material. It is actively involved in the preparation of dimethylaminoethylacrylate (DMAEA) which is used as a monomer to make flocculants for water treatment. It plays a vital role in the manufacture of water-based latex paints and adhesives, textile and paper coatings and leather finish resins.

Solubility
Miscible with ethanol, ethyl ether and chloroform. Silghtly miscible with water and dimethyl sulfoxide.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids, amines, peroxides, chlorosulfonic acid, moisture and strong alkalies.
TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 140-88-5
Formula molecolare C5H8O2
Molecular Weight (g/mol) 100.12
Numero MDL MFCD00009188
InChI Key JIGUQPWFLRLWPJ-UHFFFAOYSA-N
Sinonimo ethyl acrylate, acrylic acid ethyl ester, ethyl propenoate, 2-propenoic acid, ethyl ester, ethyl 2-propenoate, ethylacrylaat, ethylakrylat, etil acrilato, acrylic acid, ethyl ester, aethylacrylat
PubChem CID 8821
ChEBI CHEBI:82327
IUPAC Name ethyl prop-2-enoate
SMILES CCOC(=O)C=C

Specifica

Materiale o nome chimico Ethyl acrylate
Nota nome stabilized with ≈20ppm 4-methoxyphenol
Punto di fusione -71°C
Densità 0.924
Punti di ebollizione 99°C to 100°C
Punto d'infiammabilità −1°C (30°F)
Odore Pungent
Formula lineare CH2=CHCO2CH2CH3
Indice di rifrazione 1.406
Quantità 500 mL
Numero UN UN1917
Beilstein 773866
Indice di Merck 14,3759
Informazioni di solubilità Miscible with ethanol,ethyl ether and chloroform. Silghtly miscible with water and dimethyl sulfoxide.
Peso formulazione 100.12
Percent Purity 99%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.