missing translation for 'onlineSavingsMsg'
Learn More

Ethyl caprate, 99+%, Thermo Scientific Chemicals

Catalog Number p-128642
Click to view available options
Quantità:
5 mL
100 mL
500 mL
2.5 L
Imballaggio:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
110-38-3
C12H24O2
200.32
MFCD00009581
RGXWDWUGBIJHDO-UHFFFAOYSA-N
ethyl caprate, ethyl caprinate, ethyl decylate, decanoic acid ethyl ester, decanoic acid, ethyl ester, capric acid ethyl ester, ethyl n-decanoate, capric acid, ethyl ester, natural, unii-gy39fb86uo
8048
CHEBI:87430
ethyl decanoate
CCCCCCCCCC(=O)OCC
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
110-38-3
C12H24O2
200.32
MFCD00009581
RGXWDWUGBIJHDO-UHFFFAOYSA-N
ethyl caprate, ethyl caprinate, ethyl decylate, decanoic acid ethyl ester, decanoic acid, ethyl ester, capric acid ethyl ester, ethyl n-decanoate, capric acid, ethyl ester, natural, unii-gy39fb86uo
8048
CHEBI:87430
ethyl decanoate
CCCCCCCCCC(=O)OCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico Ethyl caprate
Nota nome 99+%
Acidità 1mg KOH/g max.
Punto di fusione -20.0°C
Colore Colorless
Densità 0.8665g/mL
Punti di ebollizione 245.0°C
Punto d'infiammabilità 102°C
Spettro a infrarossi Authentic
Intervallo percentuale saggio 99% min. (GC)
Imballaggio Glass bottle
Formula lineare CH3(CH2)8CO2C2H5
Indice di rifrazione 1.4240 to 1.4270
Quantità 5 mL
Beilstein 02,356
Indice di Merck 15,3831
Gravità specifica 0.8665
Informazioni di solubilità Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether
Viscosità <30 mPa.s (20°C)
Peso formulazione 200.32
Percent Purity 99+%
Forma fisica Liquid
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'