missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl Chrysanthemate, TRC
Specifications
Specifications
| Materiale o nome chimico | Ethyl Chrysanthemate |
| CAS | 97-41-6 |
| Formula molecolare | C12H20O2 |
| Quantità | 25 g |
| Sinonimo | 2,2-Dimethyl-3-(2-methyl-1-propen-1-yl)cyclopropanecarboxylic Acid Ethyl Ester, Chrysanthemummonocarboxylic Acid Ethyl Ester, 2,2-dimethyl-3-(2-methylpropenyl)cyclopropanecarboxylic Acid Ethyl Ester, Chrysanthemic Acid Ethyl Ester, Chrysanthemumic Acid Ethyl Ester, Ethyl 2,2-Dimethyl-3-(2-methyl-1-propenyl)cyclopropanecarboxylate, Ethyl chrysanthemumate, NSC 10819 |
| InChI Formula | InChI=1S/C12H20O2/c1-6-14-11(13)10-9(7-8(2)3)12(10,4)5/h7,9-10H,6H2,1-5H3 |
| SMILES | CCOC(=O)C1C(C=C(C)C)C1(C)C |
| Molecular Weight (g/mol) | 196.29 |
| Peso formulazione | 196.1463 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?