missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Isobutyrate, TRC

Codice prodotto. 30384707
Cambia vista
Click to view available options
Quantità:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
30384707 100 g
30373791 25 g
30371198 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30384707 Fornitore Toronto Research Chemicals N. del fornitore TRCE921305100G

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 97-62-1
Formula molecolare C6 H12 O2
Molecular Weight (g/mol) 116.16
Sinonimo Isobutyric acid, ethyl ester (6CI, 8CI), 2-Methylpropanoic acid ethyl ester, 2-Methylpropionic acid ethyl ester, Ethyl 2,2-dimethylacetate, Ethyl 2-methyl-1-propanoate, Ethyl 2-methylpropanoate, Ethyl 2-methylpropionate, Ethyl isobutanoate, Ethyl isobutyrate, NSC 97194
IUPAC Name ethyl 2-methylpropanoate
SMILES CCOC(=O)C(C)C

Specifica

Materiale o nome chimico Ethylisobutyrate
Quantità 100 g
InChI Formula InChI=1S/C6H12O2/c1-4-8-6(7)5(2)3/h5H,4H2,1-3H3
Peso formulazione 116.0837
Titolo del prodotto
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.