Learn More
Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate, 97%, ee 98+%
Codice prodotto. 11339126
Cambia vista
Click to view available options
Quantità:
5 g
25 g
Dimensione della confezione:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11339126 5 g 5g
11349126 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11339126 Fornitore Thermo Scientific Alfa Aesar N. del fornitore L20259.06
Richiesta di formato massivo o personalizzato
CAS: 312745-91-8 | C7H11NO3 | 157.17 g/mol
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.
Solubility
Not miscible or difficult to mix with water.
Notes
Store away from water/moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 312745-91-8
Formula molecolare C7H11NO3
Molecular Weight (g/mol) 157.17
Numero MDL MFCD00672878
InChI Key LOQFROBMBSKWQY-UHFFFAOYNA-N
Sinonimo ethyl s-4-cyano-3-hydroxybutyrate, s-ethyl 4-cyano-3-hydroxybutanoate, ethyl 3s-4-cyano-3-hydroxybutanoate, s-ethyl-4-cyano-3-hydroxybutyrate, ethyl s-+-4-cyano-3-hydroxybutyrate, butanoic acid, 4-cyano-3-hydroxy-, ethyl ester, 3s, 4-cyano-3-hydroxy-butyric acid ethyl ester, pubchem21112
PubChem CID 7020868
IUPAC Name ethyl (3S)-4-cyano-3-hydroxybutanoate
SMILES CCOC(=O)CC(O)CC#N
Specifica
Materiale o nome chimico Ethyl (S)-(+)-4-cyano-3-hydroxybutyrate
Densità 1.114
Punti di ebollizione 270°C
Punto d'infiammabilità >110°C (230°F)
Note percentuale saggio ee ≥98%
Indice di rifrazione 1.448
Quantità 5 g
Informazioni di solubilità Not miscible or difficult to mix with water.
Optical Rotation +5° (Neat)
Peso formulazione 157.17
Percent Purity 97%