missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylene Terephthalate Cyclic Trimer, TRC
Specifica
Specifica
| Materiale o nome chimico | Ethylene Terephthalate Cyclic Trimer |
| Note sul grado di purezza | HPLC |
| Quantità | 5 mg |
| InChI Formula | InChI=1S/C30H24O12/c31-25-19-1-2-20(4-3-19)26(32)38-15-16-40-28(34)22-9-11-24(12-10-22)30(36)42-18-17-41-29(35)23-7-5-21(6-8-23)27(33)39-14-13-37-25/h1-12H,13-18H2 |
| Percent Purity | >95% |
Titolo del prodotto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?