Learn More
Thermo Scientific Chemicals Ethylenediamine-N,N'-diacetic acid
Codice prodotto. 15415769
Cambia vista
Click to view available options
Quantità:
5 g
25 g
Dimensione della confezione:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
15415769 5 g 5g
15425769 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 15415769 Fornitore Thermo Scientific Chemicals N. del fornitore J63570.06
Richiesta di formato massivo o personalizzato
Acts as a chelating agent | CAS: 5657-17-0 | C6H10N2Na2O4 | 220.14 g/mol
Ethylenediamine-N,N'-diacetic acid is acts as a chelating agent, used as a haemostatic.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Ethylenediamine-N,N′-diacetic acid is acts as a chelating agent, used as a haemostatic.
Solubility
Soluble in warm water.
Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 5657-17-0
Formula molecolare C6H10N2Na2O4
Molecular Weight (g/mol) 220.14
Numero MDL MFCD00004281
InChI Key QMIFISRIAJBCGZ-UHFFFAOYSA-L
Sinonimo edda, ethylenediamine-n,n'-diacetic acid, n,n'-ethylenediglycine, glycine, n,n'-1,2-ethanediylbis, ethylenediaminediacetic acid, glycine, n,n'-ethylenedi, unii-4lx0u5342u, 2-2-carboxymethylamino ethylamino acetic acid, ethylenediamine-n,n'-diaceticacid, ethylenediamine-n,n'-di acetic acid
PubChem CID 61975
SMILES [Na+].[Na+].[O-]C(=O)CNCCNCC([O-])=O
Specifica
Materiale o nome chimico Ethylenediamine-N,N'-diacetic acid
Punto di fusione 224°C
Colore White
Formula lineare HOOCCH2NHCH2CH2NHCH2COOH
Quantità 5 g
Beilstein 1778355
Informazioni di solubilità Soluble in warm water.
IUPAC Name 2-[2-(carboxymethylamino)ethylamino]acetic acid
Peso formulazione 176.17
Forma fisica Powder