Learn More
Thermo Scientific™ Eutech Handheld Meters Calibration Solution, 84μ S / cm KCl
Product Code. 15264878
Change view
Click to view available options
Quantity:
480 mL
Unit Size:
480mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15264878 480 mL 480mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15264878 Supplier Thermo Scientific™ Supplier No. 01X211241
  • ONLINE EXCLUSIVE PRICE
    18.35€
    17.30€ / 480mL
    Risparmia 1.05€
    5% Sconto
 Restricted Item
In stock
Add to basket
Edge
Add to basket
  • 84μS/cm KCl calibration solution
  • Solutions packed in 480mL durable plastic bottles
  • Economical and convenient for laboratory use.
TRUSTED_SUSTAINABILITY
Specifications
Odor Odorless
Density 1.0 g/cm³ at 25°C
Physical Form Liquid
pH ∼7
Boiling Point ∼100°C
For Use With (Equipment) Eutech Handheld Meters
Solution Type Calibration Solution
Type Calibration Solution
Industry Type Laboratory Use
Quantity 480 mL