missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eutech™ ECFG7370101B pH electrode

Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. 

Marca:  Thermo Scientific™

Dettagli aggiuntivi : Peso : 0.12600kg

Codice prodotto. 16356395

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi »


Descrizione

Descrizione

The measure of a liquid acidity and alkalinity, pH is one of the most common parameter measured in a wide variety of industries ranging from water and wastewater treatment, chemical production, agriculture research and production, environmental monitoring, chemical and life sciences research, biochemical and pharmaceutical research, electronics production to food processing and other industrial applications. Eutech™ offers a wide range of pH meters and electrodes that measure pH values in various educational, laboratory and industrial applications. This clog free double-junction glass pH electrode is ideal for samples containing TRIS buffers, sulfides and more. Easy to operate fill hole sleeve mechanism is used for general pH measurements. Available with 1 m cable.

Specifications

  • Parameter: pH
  • Range: 0 to 13 pH
  • Temperature Range: 0 to 100 °C
  • Liquid Junction Type: Annular ceramic
  • Internal Reference Type: Ag/AgCl
  • Sealed/Refillable: Refillable
  • Reference Junction: Double
  • Refilling Reference Electrolyte: ECRE002
  • Dimensions (Shaft): 110 x 12mm
  • Cable Length: 1m
  • Connector: BNC
  • Used With: All pHmeters with BNC input connector
  • Specifica

    Specifica

    BNC
    0 to 13
    pH Electrode
    Double
    0°C to 100°C
    Suggerimenti di prodotto

    Suggerimenti di prodotto

    Videos
    SDS
    Documenti

    Documenti

    Certificati
    Offerte speciali

    Offerte speciali