Learn More
Eutech™ KCl Calibration Solution
Product Code. 15223657
Change view
Click to view available options
Quantity:
60 mL
480 mL
Conductivity:
100 μS/cm
111.8 mS/cm
12.88 mS/cm
1413 μS/cm
2764 μS/cm
500 μS/cm
84 μS/cm
Unit Size:
480mL
500mL
60mL
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Conductivity Unit Size
15223657 60 mL 84 μS/cm 60mL
18854276 480 mL 84 μS/cm -
12703626 480 mL 100 μS/cm 500mL
12753626 480 mL 500 μS/cm 500mL
10775193 480 mL 1413 μS/cm 500mL
17266033 60 mL 1413 μS/cm 60mL
15386487 480 mL 2764 μS/cm 480mL
15488972 480 mL 12.88 mS/cm 480mL
10018182 480 mL 12.88 mS/cm 500mL
17276033 60 mL 12.88 mS/cm 60mL
10602614 480 mL 111.8 mS/cm 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15223657 Supplier Thermo Scientific Eutech Supplier No. 01X609201
  • ONLINE EXCLUSIVE PRICE
    10.80€
    9.00€ / 60mL
    Risparmia 1.80€
    16% Sconto
 Restricted Item
In stock
Add to basket
Edge
Add to basket
For conductivity calibration and standards, choose from our available Eutech™'s Conductivity Standards, Calibrations, and Solutions
Eutech™ KCl Calibration Solutions are used for calibrating conductivity probes for accurate measurement. Get quick and accurate conductivity measurements using Eutech™ conductivity meters and probes in the field, laboratories, and schools.
Features:
  • Packed in 60mL or 480 ml durable plastic bottles 
  • Economical and convenient to use
TRUSTED_SUSTAINABILITY
Specifications
Description 84 μS/cm KCl Calibration Solution
Conductivity 84 μS/cm
For Use With (Equipment) Conductivity Meters and Electrodes
Solution Type Calibration Solution
Type Calibration Solution
Quantity 60 mL