Learn More
Lenz Laborglasintrumente Evaporating Flasks with Conical Ground Joint
Codice prodotto. 11953381
Cambia vista
Click to view available options
Lato giunto rastremato standard:
NS 14/23
NS 29/32
Dimensione della confezione:
1 PCS
pezzi 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Lato giunto rastremato standard Dimensione della confezione
11953381 NS 14/23 pezzi 10
11886104 NS 29/32 1 PCS
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11953381 Fornitore Lenz Laborglasintrumente N. del fornitore 03041410
  • ONLINE EXCLUSIVE PRICE
    201.00€
    137.60€ / pezzi 10
    Risparmia 63.40€
    31% Sconto
Spedizione stimata: 04-01-2027
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Especially for rotary evaporators
  • With center neck and conical joint
  • Pear-shaped
  • With inscription label
  • Material: DURAN™
Specifica
Materiale Duran Glass
Lato giunto rastremato standard NS 14/23
Volume (sistema metrico) 5 mL