Learn More
Lenz Laborglasintrumente Evaporating Flasks with Conical Ground Joint, with PUR Coating
Codice prodotto. 11903391
Cambia vista
Click to view available options
Volume (sistema metrico):
50 mL
100 mL
250 mL
2 L
3 L
Dimensione della confezione:
Pezzo
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Volume (sistema metrico) unitSize
11903391 100 mL Pezzo
15695585 250 mL Pezzo
11943391 2 L Pezzo
11953391 3 L Pezzo
11993381 50 mL Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Codice prodotto. 11903391 Fornitore Lenz Laborglasintrumente N. del fornitore 03044037
  • ONLINE EXCLUSIVE PRICE
    100.00€
    65.21€ / pezzi 1
    Risparmia 34.79€
    34% Sconto
Spedizione stimata: 31-08-2026
Aggiungi al carrello
Aggiungi al carrello
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Especially for rotary evaporators
  • With center neck and conical joint
  • With highly transparent PUR coating
  • Autoclavable
  • Pear-shaped
  • Material: DURAN™
Specifica
Materiale Duran Glass
Volume (sistema metrico) 100 mL
Diametro (sistema metrico) 60 mm