Learn More
Liebisch™ Evaporator with Fixed Heating Block
Codice prodotto. 10118191
Cambia vista
Click to view available options
Temperature Range (Metric):
Ambient to 200°C
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Temperature Range (Metric) Dimensione della confezione
10118191 Ambient to 200°C Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 10118191 Fornitore Liebisch™ N. del fornitore 67186101
 Articolo soggetto a restrizioni
Spedizione stimata: 10-12-2026
Metal block thermostat combined with a patented multiplex valve depot for superior evaporation operations. Liebisch™ Evaporator with Fixed Heating Block can evaporate various samples under targeted aeration.
  • Metal block thermostat / height-adjustable multiplex valve cepot
  • Interchangeable stainless steel luer needles with plug-in connections.
  • valve depot can be swiveled sideways for convenient sample loading
Specifica
Tensione 230 V
Temperature Range (Metric) Ambient to 200°C
Dimensioni (L x P x A) 455 x 230 x 130 mm