Learn More
R&D Systems™ Everolimus
Product Code. 18731598
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18731598 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18731598 Supplier R&D Systems™ Supplier No. 6188/10
  • ONLINE EXCLUSIVE PRICE
    332.89€
    291.90€ / 10mg
    Risparmia 40.99€
    12% Sconto
 Restricted Item
Spedizione stimata: 19-11-2026
A Tocris product, part of R&D Systems. mTOR inhibitor;immunosuppressant
Everolimus is a mTOR inhibitor. Inhibits tumor proliferation in vitro and in vivo. Inhibits VEGF-induced HUVEC proliferation in vitro, but does not affect tumor vascular permeability in mouse models. Immunosuppressant. Activates autophagy.
TRUSTED_SUSTAINABILITY
Specifications
CAS 159351-69-6
Quantity 10 mg
Product Line Tocris
Target mTOR
Molecular Formula C53H83NO14
Formula Weight 958.22g/mol
Product Type Inhibitor
For Research Use Only