Learn More
Safety First Aid Evolution Minibus And Bus First Aid Kit, BS8599-2 Compliant
Descrizione
- Compliant with British Standard BS8599-2 requirements for Motor Vehicles.
- Ideal for minibuses and small buses.
- Buses and coaches that transport more than 17 passengers require 2 large vehicle kits.
- A risk assessment should be carried out to identify if extra first aid supplies are needed.
- Dimensions: 30 cm x 25 cm x 12 cm
- The BS8599-2 Minibus and Bus First Aid Kit in Evolution Case Contains:
- 1 x First Aid Guidance Leaflet
- 2 x HypaCover Trauma Dressings, Medium
- 1 x HypaCover Trauma Dressing, Large
- 2 x HypaCover Sterile Dressings, 12 x 12 cm
- 2 x HypaBand Triangular Bandages
- 2 x HypaCover Adhesive Dressings, 10 x 8 cm
- 20 x HypaPlast Washproof Plasters
- 5 x HypaTouch Nitrile Gloves
- 2 x HypaGuard Face Shields
- 3 x HypaGuard Foil Blankets
- 20 x HypaClean Sterile Wipes
- 4 x HypaSoothe Burns Dressings, 10 x 10 cm
- 1 x Clothing Cutters
Specifica
Specifica
| Product Type | First Aid Kit |
| Materiale | Plastic |
| Sterilità | Sterile |