Learn More
R&D Systems™ ExCellerate NK Cell Expansion Media, Animal Component-Free
Specifica
Specifica
| Product Line | R&D Systems |
| Quantity | 1000 mL |
| Regulatory Status | Animal Free |
| Species | Human |
| Content And Storage | Store the unopened product at -20°C to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |