Learn More
Epredia™ Excelsior™ AS Tissue Processor Accessories
Codice prodotto. 12648156 Sfoglia Tutto Epredia Prodotti
Cambia vista
Click to view available options
Product Type:
Air System Duct Adapter Kit
Basket Carrying Container
Downdraft Extrctn Duct Adapter
Filtro a potassio permanganato per la rimozione della formaldeide
Monitor Clamp Kit
Organized Basket, 222 cassette
Organized Basket, 300 cassette
Storage Bottle
Vapor Filter (Charcoal)
Waste Wax Drawer Kit
Dimensione della confezione:
Pezzo
pezzi 6
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Product Type unitSize
12648156 Air System Duct Adapter Kit Pezzo
12638156 Downdraft Extrctn Duct Adapter Pezzo
12683156 Storage Bottle pezzi 6
12678156 Waste Wax Drawer Kit Pezzo
12688156 Monitor Clamp Kit Pezzo
12608346 Basket Carrying Container Pezzo
13470487 Vapor Filter (Charcoal) pezzi 6
12764856 Filtro a potassio permanganato per la rimozione della formaldeide pezzi 6
12658156 Organized Basket, 222 cassette pezzi 6
15594280 Organized Basket, 300 cassette Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Codice prodotto. 12648156 Fornitore Epredia™ N. del fornitore A78410024
  • ONLINE EXCLUSIVE PRICE
    171.00€
    146.91€ / pezzi 1
    Risparmia 24.09€
    14% Sconto
Spedizione stimata: 04-09-2026
Aggiungi al carrello
Aggiungi al carrello
Maintain the Epredia™ Excelsior™ AS Tissue Processor with Epredia™ Excelsior AS Tissue Processor Accessories.
For use with Shandon Excelsior ES Tissue Processor
TRUSTED_SUSTAINABILITY
Specifica
Product Type Air System Duct Adapter Kit
Da utilizzare con (apparecchiatura) Excelsior AS Tissue Processor