missing translation for 'onlineSavingsMsg'
Learn More

EXTL2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA567024

Dettagli aggiuntivi : Peso : 0.01000kg

Codice prodotto. 15909704

  • 514.00€ / 100µL
Spedizione stimata: 02-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: TANRMRNRLQV FPELETNAVL MVDDDTLIST PDLVFAFSVW QQFPDQIVGF VPRKHVSTSS GIYSYGSFEM Highest antigen sequence identity to the following orthologs - mouse 89%, rat 90%.

EXTL2 Polyclonal antibody specifically detects EXTL2 in Human samples. It is validated for Immunocytochemistry.
Specifica

Specifica

EXTL2
Polyclonal
Unconjugated
EXTL2
3000001D04Rik; alpha-1,4-N-acetylhexosaminyltransferase EXTL2; Alpha-GalNAcT EXTL2; AW146439; exostoses (multiple)-like 2; exostoses-like 2; exostosin like glycosyltransferase 2; exostosin-like 2; exostosin-like glycosyltransferase 2; exotoses (multiple)-like 2; EXTL2; Extr2; EXT-related protein 2; Glucuronyl-galactosyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase; processed exostosin-like 2
Rabbit
Antigen affinity chromatography
RUO
2135
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9UBQ6
EXTL2
Recombinant Human EXTL2
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

For Research Use Only