Learn More
Macherey Nagel Extraction Thimbles, Cellulose, MN 645
Often used for holding solid material from which certain substances are to be eluted (extracted) with a suitable solvent.
Marca:  Macherey Nagel 645039
Dettagli aggiuntivi : Peso : 0.10000kg
Codice prodotto. 10236211
  • 209.00€ / pezzi 25
Spedizione stimata
15-03-2021
Aggiungi al carrello
Descrizione
Descrizione
  • Often used for holding solid materials from which certain substances are to be eluted (extracted) with a suitable solvent
  • Used in the fields of air and waste gas analysis for collecting solid particles (dust)
  • Standard grade
  • Made from pure cellulose
Specifica
Specifica
60 mm
MN 645
2.0 mm
Holding
180 mm
Cellulose