Descrizione
Increase productivity by enabling to grow more cells faster and easier. 3- or 5-layer formats provide 525 cm2 and 875 cm2 cell growth surface area, respectively.
- Even distribution of media across all layers for homogeneous cell growth
- Ability to mix cells and reagents in the Falcon Multi-Flask saves time
- Flexible design allows to pour or aspirate/recover cells using a pipet
- Consistent surface treatment for predictable scale-up
- Lot number printed on each flask for traceability
- Manufactured in compliance to cGMP standards
Specifica
Specifica
| Materiale | Polistirene |
| Stile collo | Dritto |
| N. per custodia | 12/Cs. |
| N. per confezione | 2/Pk. |
| Product Type | Cell Culture Multi-Flask |
| Forma | Rettangolare |
| Rivestimento superficiale | Trattamento per coltura di tessuti su 3 strati |
| Monouso | Single-use |
| Graduated | Yes |
| Sterilità | Sterili |
| Vedi altri risultati |
Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.