Includes
24-well insert plate, 24-well plate or single-well feeder tray, and lid
Falcon 24 Multiwell Insert Systems are designed to automate many commonly used cell-based assays for drug discovery. Available in a choice of membrane pore sizes, 24 Multiwell inserts have been successfully used for a variety of applications including permeability studies for oral bioavailability (e.g., Caco-2 cells), chemotaxis, cell migration and invasion assays. These insert systems offer all the benefits of Falcon Individual Cell Culture Inserts in an automation-friendly format that is compatible with most robots and fluid handlers. The Falcon 24 Multiwell Insert Plate is a single unit that is compatible with all Falcon 24 well plates and Feeder Tray.
Specifica
Specifica
| Colore | Trasparente |
| Area di coltura | 0.3 cm2 |
| Materiale | PET |
| Sterilità | Sterile |
| Trattamento superficiale | Cell Culture |
| Tipo | Multi-well Insert System |
| Conteggio pozzetti | 24 |
| Da utilizzare con (applicazione) | Eccellente per studi di differenziazione, tossicologia, migrazione e invasione |
| Include | Piastra a inserti da 24 pozzetti, vassoio di alimentazione a pozzetto singolo e coperchio |
| N. per custodia | 5/Cs. |
| Vedi altri risultati |