missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FAM18B Polyclonal Antibody

Codice prodotto. 13210159
Cambia vista
Click to view available options
Quantità:
100 μL
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
13210159 100 μL 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 13210159 Fornitore Invitrogen™ N. del fornitore PA521971

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Rabbit Polyclonal Antibody

Recommended positive controls: HeLa. Predicted reactivity: Rhesus Monkey (100%), Chimpanzee (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The FAM18B gene (Family with Sequence Similarity 18 Member B) is located on chromosome 17q21.31 and encodes a protein whose precise biological functions are not fully characterized. The FAM18 family, to which FAM18B belongs, includes proteins that are presumed to be involved in cellular processes such as signal transduction and intracellular trafficking. Although the specific role of FAM18B remains under investigation, initial studies suggest it may be involved in modulating cellular responses to environmental stimuli and maintaining cellular homeostasis. There is limited research on FAM18B's involvement in disease, but its expression patterns in various tissues, including the brain and reproductive organs, indicate potential roles in neurological and reproductive functions.
TRUSTED_SUSTAINABILITY

Specifica

Antigene FAM18B
Applicazioni Western Blot
Classificazione Polyclonal
Concentrazione 1 mg/mL
Coniugato Unconjugated
Formulazione 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene TVP23B
N. accesso geni Q9NYZ1
Alias gene CGI-148; FAM18B; FAM18B1; family with sequence similarity 18, member B; family with sequence similarity 18, member B1; Golgi apparatus membrane protein TVP23 homolog B; NPD008; protein FAM18B1; trans-golgi network vesicle protein 23 homolog B (S. cerevisiae); TVP23B; YDR084C
Simboli geni TVP23B
Specie ospite Rabbit
Immunogeno Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of human FAM18B. The exact sequence is proprietary.
Metodo di purificazione Antigen affinity chromatography
Quantità 100 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 51030
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Forma Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.