missing translation for 'onlineSavingsMsg'
Learn More

FAM69A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Thermo Scientific PA553573

Dettagli aggiuntivi : Peso : 0.02000kg

Codice prodotto. 15741511

  • 514.00€ / 100µL
Spedizione stimata: 11-04-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Immunogen sequence: IICDKYKTGV IDGPACNSLC VTETLYFGKC LSTKPNNQMY LGIWDNLPGV VKCQMEQALH LDFGTELEPR KEIVLFDKPT RGTTVQKFKE MVYSLFKAKL GDQGNLSELV NLILTVADGD KDGQVSLGEA KSAWA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

FAM69A Polyclonal antibody specifically detects FAM69A in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifica

Specifica

FAM69A
Polyclonal
Unconjugated
FAM69A
DIPK1A; Divergent protein kinase domain 1A; FAM69A; family with sequence similarity 69 member A; family with sequence similarity 69, member A; protein FAM69A
Rabbit
Antigen affinity chromatography
RUO
388650
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q5T7M9
FAM69A
Recombinant protein corresponding to Human FAM69A
100 μL
Primary
Human
Antibody
IgG
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali

Research Use Only