Learn More
3M™ Fastener Hook & Loop SJ352B
Product Code. 17207800 Shop All 3M Products
Change view
Click to view available options
Dimensions (L x W):
1.25 m x 25 mm
Unit Size:
4 rolls
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
17207800 1.25 m x 25 mm 4 rolls
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17207800 Supplier 3M™ Supplier No. 7100123266
  • ONLINE EXCLUSIVE PRICE
    105.00€
    59.00€ / 4 rotoli
    Risparmia 46.00€
    43% Sconto
Spedizione stimata: 11-12-2026
Fastener Hook & Loop
  • High performance rubber based adhesive designed for indoor applications requiring medium temperature resistance
  • Provides a firm adhesive bond to a wide variety of surfaces
  • Woven nylon is durable for thousands of closures
  • Quick fastening that helps reduce production time and costs
TRUSTED_SUSTAINABILITY
Specifications
Product Type Fastener
Format Roll
Color Black
Dimensions (L x W) 1.25 m x 25 mm
Thickness (Metric) 4.4 mm