Learn More
Feed Kit A+, B+, C+
Specifica
Specifica
| Content And Storage | • 1 x 1 L EfficientFeed A+ Supplement • 1 x 1 L EfficientFeed B+ Supplement • 1 x 1 L EfficientFeed C+ Supplement Store above in refrigerator (2–8°C). Protect from light and moisture. • Flash drive with manual |
| Linea cellulare | CHO |
| Classificazione | Chemically-defined |
| Forma | AGT |
| Product Type | Media Supplement |
| Quantità | 3 L |