Learn More
Value Plastics Female Luer to 500 Series Barb
Codice prodotto. 30053447 Sfoglia Tutto Cole-Parmer Prodotti
Cambia vista
Click to view available options
Ingresso:
Female Luer x Hose Barb
Dimensione della confezione:
pezzi 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Ingresso Dimensione della confezione
30053447 Female Luer x Hose Barb pezzi 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30053447 Fornitore Value Plastics N. del fornitore FTLLSB0136005
  • ONLINE EXCLUSIVE PRICE
    23.85€
    21.52€ / pezzi 25
    Risparmia 2.33€
    9% Sconto
Spedizione stimata: 18-12-2026
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifica
Certificazione/Conformità ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Ingresso Female Luer x Hose Barb
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.