Learn More
Value Plastics Female Luer Thread Style to UNF Threaded Adapter Fittings, Straight
Codice prodotto. 30053983 Sfoglia Tutto Cole-Parmer Prodotti
Cambia vista
Click to view available options
Dimensioni filo:
1/4-28 UNF(M)
1/4-28 UNF(M) Bottom Sealing
10-32 UNF(M)
Ingresso:
Female Luer with 1/4 in. Hex
Female Luer with 5/16 in. Hex
Materiale (guarnizione):
Nylon
Polycarbonate
Polypropylene
Dimensione della confezione:
pezzi 25
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Dimensioni filo Ingresso Materiale (guarnizione) Dimensione della confezione
30053983 1/4-28 UNF(M) Female Luer with 5/16 in. Hex Polypropylene pezzi 25
30053201 1/4-28 UNF(M) Bottom Sealing Female Luer with 5/16 in. Hex Polypropylene pezzi 25
30053683 10-32 UNF(M) Female Luer with 1/4 in. Hex Nylon pezzi 25
30054148 1/4-28 UNF(M) Bottom Sealing Female Luer with 5/16 in. Hex Nylon pezzi 25
30053992 1/4-28 UNF(M) Female Luer with 5/16 in. Hex Nylon pezzi 25
30054264 1/4-28 UNF(M) Female Luer with 5/16 in. Hex Polycarbonate pezzi 25
30054098 10-32 UNF(M) Female Luer with 1/4 in. Hex Polycarbonate pezzi 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Codice prodotto. 30053983 Fornitore Value Plastics N. del fornitore SFTLL6005
  • ONLINE EXCLUSIVE PRICE
    52.55€
    47.04€ / pezzi 25
    Risparmia 5.51€
    10% Sconto
Spedizione stimata: 06-11-2026
Luer fittings allow for a simple yet secure connection
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Specifica
Certificazione/Conformità ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Quantità confezione 25/Pk.
Ingresso Female Luer with 5/16 in. Hex
Dimensioni filo 1/4-28 UNF(M)
Materiale (guarnizione) Polypropylene