Descrizione
Quick and easy assembly and disassembly without the need for tools. Thread design provides a secure closure—preventing contamination. Conforms to ISO-594-1 and ISO 594-2 standards
Specifica
Specifica
| Certificazione/Conformità | Non-DEHP;Non-Phthalate;RoHS;FDA |
| Quantità confezione | 25/Pk. |
| Ingresso | Female Luer Thread |
| Materiale (guarnizione) | Polycarbonate |