Descrizione
- Dispenses accurate volumes directly from drums and bulk refills.
- Remote system allows storage of the drum up to 10 meters (30 feet) away from the Dispensette™.
- Max. delivery height is approximately 1.2m.
- For use with Dispensette™III/Organic
Specifica
Specifica
| Product Type | Remote Dispensing System |
| Da utilizzare con (apparecchiatura) | Dispensette™III/Organic |
| Include | Plug-in adapter (PTFE, only for Dispensette ≤ 10 ml), Thread adapter (PP, GL 45/32), Stain-free grip (PP), Locking screw (ETFE), Wall mounting unit ( PP), Thread adapter (PP, GL 32/28), Filling tube (FEP, 3 m, outer Ø 7.6mm), Mounting screw (PTFE), Locking screw (ETFE), Coupling (ETFE, with ball valve), Drum adapter (PTFE, for drums with inner-thread of 3/4 in., with ball valve, incl. closure cap), Membrane filter (3 μm, non-sterile), Filling tube (470mm, outer Ø 6.9mm) |
| Quantità | 1 Ea. |