Description
GeneRuler 100bp 100 to 1000bpDNA Ladder is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler 100bp DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol).
The ladder is supplied with 6X DNA Loading Dye.
- Ideal for both DNA sizing and approximate quantification
- Sharp bands
- Bright reference bands (given in red)
- Supplied with loading dye for sample DNA
Specifications
Specifications
| Concentrazione | 6X |
| Gel Type | Agarose |
| Label or Dye | Xylene Cyanol FF, Bromophenol Blue, Orange G |
| Ready to Load | No |
| Intervallo dimensioni | 100 to 1000 bp |
| Compatibilità gel | Agarose Gels |
| Quantità | 50 µg |
| Product Line | GeneRuler |
| Product Type | DNA Ladder |