Learn More
Thermo Scientific™ Scala DNA GeneRuler™ 100bp
Codice prodotto. 11873953 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
5 x 50 µg
50 µg
Dimensione della confezione:
50µg
pezzi 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
11873953 5 x 50 µg pezzi 5
10588170 50 µg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 11873953 Fornitore Thermo Scientific™ N. del fornitore SM0242
  • ONLINE EXCLUSIVE PRICE
    292.00€
    259.65€ / pezzi 5
    Risparmia 32.35€
    11% Sconto
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
In stock
Aggiungi al carrello
Edge
Aggiungi al carrello
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.
GeneRuler 100bp 100 to 1000bpDNA Ladder is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler 100bp DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol).
The ladder is supplied with 6X DNA Loading Dye.
The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler 100 bp DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase, protocol. The ladder is supplied with 6X DNA Loading Dye.
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY
Specifica
Concentrazione 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Ready to Load No
Intervallo dimensioni 100 to 1000 bp
Compatibilità gel Agarose Gels
Quantità 5 x 50 µg
Product Line GeneRuler
Product Type DNA Ladder