Learn More
Thermo Scientific™ Scala DNA GeneRuler™ 1 kb
Product Code. 11833963 Shop All Thermo Scientific Products
Change view
Click to view available options
Contenuto kit:
6X DNA Loading Dye, 2 x 1 ml
Colorante di caricamento DNA 6X, 10 x 1 ml
Unit Size:
pezzi 25
pezzi 5
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Contenuto kit Unit Size
11833963 Colorante di caricamento DNA 6X, 10 x 1 ml pezzi 25
11823963 6X DNA Loading Dye, 2 x 1 ml pezzi 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11833963 Supplier Thermo Scientific™ Supplier No. SM0312
  • ONLINE EXCLUSIVE PRICE
    556.00€
    483.65€ / 50µg
    Risparmia 72.35€
    13% Sconto
View all promotions
 Articolo soggetto a restrizioni
In stock
Add to basket
Edge
Add to basket
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels with this mixture of chromatography-purified individual DNA fragments.
GeneRuler 1kb DNA Ladder, range 250 to 10,000bp can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol).
The ladder is supplied with 6X DNA Loading Dye, and it can be used in DNA sizing and approximate quanitification applications.
The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler 1kb DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase. The ladder is supplied with 6X DNA Loading Dye.
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY
Specifications
Concentrazione 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Ready to Load No
Intervallo dimensioni 250 to 10000 bp
Compatibilità gel Agarose Gels
Contenuto kit Colorante di caricamento DNA 6X, 10 x 1 ml
Quantità 25 x 50 μg
Product Line GeneRuler
Product Type DNA Ladder