Learn More
Thermo Scientific™ Scala DNA 100 bp GeneRuler, pronta all'uso 100-1000 bp
Codice prodotto. 10328179 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Product Type:
100BP DNA Ladder
100BP Plus DNA Ladder
1KB DNA Ladder
1KB Plus DNA Ladder
50BP DNA Ladder
DNA Ladder Mix
High Range DNA Ladder
Low Range DNA Ladder
Ultra Low Range DNA Ladder
Quantità:
100 Applications
500 Applications
Intervallo dimensioni:
10 to 300 bp
100 to 1000 bp
100 to 3000 bp
10171 to 48502 bp
25 to 700 bp
250 to 10000 bp
50 to 1000 bp
75 to 20000 bp
Number of Markers:
8
10
11
13
14
15
21
Dimensione della confezione:
50µg
pezzi 5
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Product Type Quantità Intervallo dimensioni Number of Markers Dimensione della confezione
10328179 100BP DNA Ladder 100 Applications 100 to 1000 bp 10 50µg
10400280 Ultra Low Range DNA Ladder 100 Applications 10 to 300 bp 11 50µg
10784881 Low Range DNA Ladder 100 Applications 25 to 700 bp 10 50µg
10314340 50BP DNA Ladder 100 Applications 50 to 1000 bp 13 50µg
10600301 1KB Plus DNA Ladder 100 Applications 75 to 20000 bp 15 50µg
10364280 100BP Plus DNA Ladder 100 Applications 100 to 3000 bp 14 50µg
10181070 DNA Ladder Mix 100 Applications 100 to 1000 bp 21 50µg
10809360 1KB DNA Ladder 100 Applications 250 to 10000 bp 14 50µg
10508950 High Range DNA Ladder 100 Applications 10171 to 48502 bp 8 50µg
10101240 1KB Plus DNA Ladder 500 Applications 75 to 20000 bp 15 pezzi 5
11883953 100BP DNA Ladder 500 Applications 100 to 1000 bp 10 pezzi 5
10181260 100BP Plus DNA Ladder 500 Applications 100 to 3000 bp 14 pezzi 5
11531605 DNA Ladder Mix 500 Applications 100 to 1000 bp 21 pezzi 5
11571595 1KB DNA Ladder 500 Applications 250 to 10000 bp 14 pezzi 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
Codice prodotto. 10328179 Fornitore Thermo Scientific™ N. del fornitore SM0243
 Articolo soggetto a restrizioni
In stock
Thermo Scientific GeneRuler DNA Ladders offer a wide selection of size standard made with individual chromatography-purified fragments in the range of 10 bp to 48.5 kb.
Thermo Scientific GeneRuler DNA Ladders serve as precise molecular weight markers for the sizing and approximate quantification of double-stranded DNA fragments, accommodating a range from 10 bp up to 48.5 kb. The uniform intensity of the bands, coupled with distinct bright reference bands, facilitates accurate sizing and orientation during gel electrophoresis. Furthermore, the defined quantity of DNA within each band enables the approximate quantification of experimental DNA samples.
GeneRuler DNA ladders are provided in a convenient ready-to-use format incorporating 6X TriTrack DNA Loading Dye (contains Xylene Cyanol FF, Bromophenol Blue, and Orange G). The three dye present in the buffer allow for easy tracking of DNA migration during electrophoresis.
Highlights
  • Wide selection of DNA ladders fitting a variety of experiments
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Easy visualization—three-colored loading dye for easy tracking of DNA migration during electrophoresis
  • Ready-to-use: premixed with 6X MassRuler DNA Loading Dye for direct loading
  • Stable at room temperature for up to 24 months
Sustainability features
Ready-to-use GeneRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
TRUSTED_SUSTAINABILITY
Spezifikation
Concentrazione 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Number of Markers 10
Ready to Load Yes
Intervallo dimensioni 100 to 1000 bp
Volume (sistema metrico) 50 μg
Compatibilità gel Agarose Gels
Quantità 100 Applications
Product Line GeneRuler
Product Type 100BP DNA Ladder
Mehr anzeigen Weniger anzeigen
For Research Use Only. Not for use in diagnostic procedures.
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.