Learn More
Thermo Scientific™ Frammento di DNA NoLimits 500 bp
GREENER_CHOICE
Codice prodotto. 10611621 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Quantità:
10 μg
Dimensione della confezione:
10µg
Codice prodotto. 10611621
Marca: Thermo Scientific™ SM1641
Edge
Optimize applications where a custom DNA size standard is required with NoLimits, a collection of 36 chromatography-purified individual DNA fragments ranging from 10 -20,000bp.
NoLimits is a collection of 36 highly-purified individual DNA fragments ranging from 10 to 20,000 bp. Unlike other vendors, where custom standards are produced by PCR, Thermo Scientific NoLimits DNA fragments are produced using specifically designed plasmids purified by patented technology.
Plasmid DNA is digested with Thermo Scientific Restriction Enzymes, and the individual DNA fragments are chromatography-purified from the digestion mixture. As a result, we offer exceptionally pure DNA fragments that are free of any truncated or degraded molecules. Our custom DNA fragments are the best choice for applications, ranging from electrophoresis (both gel and capillary) to HPLC and beyond.
Highlights
  • Chromatography-purified individual DNA fragments
  • Broad range: 10 to 20,000 bp
  • Sharp peaks during capillary electrophoresis, sharp bands during gel electrophoresis
  • Precise DNA concentration 
TRUSTED_SUSTAINABILITY
Specifica
Gel Type Agarose
Product Type DNA Fragment
Ready to Load No
Intervallo dimensioni 10 to 20000 bp
Compatibilità gel Agarose Gels
Green Features Sustainable packaging
Linea di prodotti NoLimits
Quantità 10 μg