Learn More
Chemsplash FFP2 Fold Flat Mask with Valve
Descrizione
Chemsplash 3673 FFP2 Fold Flat Valved Mask
- High Filtration Capacity
- Respirator with Yellow Color Exhalation Valve
- Exhalation Valve Reduces Hot Air Buildup
- Woven Elasticated Adjustable Head Straps
- Adjustable Nose Clip with Soft Inner Nose Foam
- Folded to Pocket Size and Packed Individually for Hygiene
- Protects Against Low-Toxicity Solid and Liquid Particles
- No Vision Impairment and Problem-Free Speech
- Latex and Graphene Free
Description: The Chemsplash 3673 is an FFP2 fold-flat valved mask that comes with an exhalation valve to reduce hot air buildup. Color-coded yellow for easy identification, this mask is made of high filtration fabric, protecting against low-toxicity solid and liquid particles. A soft padded band rests inside, adjustable to ensure an excellent fit on the nose. The mask is folded to pocket size and individually packed for hygiene. It offers great comfort and ensures no vision impairment or speech difficulty. Additionally, it is latex and graphene-free for safer use.
Specifica
Specifica
| Tipo di aerosol | N - Not Resistant to Oil |
| Colore | White |
| Design Type | Flat-fold |
| Monouso | Single-use |
| Tipo di filtro | FFP2 |
| Materiale | Polypropylene |
| Product Type | General Purpose Particulate Respirator |
| Size | One Size |
| With Exhalation Valve | Sì |
| Senza lattice | Yes |
| Vedi altri risultati |