Learn More
Abnova™ FGF12/CEN3q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
Marca: Abnova™ FG0190.100uL
Dettagli aggiuntivi : Peso : 0.02000kg
Specifica
Store at 4°C in the dark. | |
Tube | |
2257 | |
100 μL |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
FISH-Ce | |
DAPI Counterstain (1500 ng/mL ) 125 uL for each 100 uL FISH Probe | |
Liquid |