Learn More
Thermo Scientific™ Fiberlite F12-6 x 500 LEX Fixed Angle Rotor with Auto-Lock for LYNX 4000 and 6000 Superspeed Centrifuges
Product Code. 13426093
Change view
Click to view available options
Capacity:
6 x 500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
13426093 6 x 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13426093 Supplier Thermo Scientific™ Supplier No. 096062375
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
 Restricted Item
Spedizione stimata: 30-11-2026
Shorten run set-up time while providing peace of mind that the rotor is secure with this advanced centrifuge rotor.
Shorten run set-up time while providing peace-of-mind that the rotor is secure with the Thermo Scientific™ Fiberlite™ F12-6 x 500 Fixed-Angle Rotor, which runs popular 500mL bottles to over 24,400 x g, and supports smaller bottles with available adapters.
Thermo Scientific™ Auto-Lock™ Rotor Exchange
  • Secure, push-button rotor exchange in less than 3 seconds
  • Improved safety and convenience
  • No tools required
  • Quickly change rotors and applications
Thermo Scientific™ Auto-ID™ Instant Rotor Identification
  • Immediate identification of a rotor
  • Rotor specifications automatically loaded into centrifuge parameters
  • Shortens run set-up time, improves safety
Speed Handle
  • Accelerates and simplifies rotor lid tightening, ensuring lid is properly attached
  • Easier and safer lifting and carrying of rotors, further enhanced with the lightweight design
TRUSTED_SUSTAINABILITY
Specifications
Description Fiberlite F12-6x500 LEX Fixed Angle Rotor
Material Carbon Fiber Composite
Type Fixed Angle Rotor
Capacity 6 x 500 mL
Max. RCF 24,471 x g
Max. Speed 12,000 rpm
Radius (Metric) 152 mm
Radius (Metric) Min. 41 mm
K Factor 1,388
Max. Tube Length 160 mm
Max. Tube Diameter 70 mm
Net Weight 20.0 lb. (9.1 kg)
For Use With (Equipment) Sorvall LYNX 6000 and 4000 Superspeed Centrifuges
Angle 20°
Biocontainment Yes
Show More Show Less
How long will the delivery take?
It is location dependent. Delivery times typically range from 2-8 weeks depending on product availability, customization requirements, and your location. Contact your sales representative for current lead times.
What is the warranty on Fiberlite rotors and metal rotors used with LYNX centrifuges?
Metal rotors 10 years, Fiberlite carbon fiber rotors 15 years
Where can I find chemical resistance information for the rotors and accessories?
Chemical resistance charts are included in the user manual and additional information is available through technical support.
How can I know what tubes can be used in my LYNX rotors?
The rotor guide of LYNX centrifuges is available on the product page under #documents