Learn More
Thermo Scientific™ Fiberlite F37L-8 x 100 Fixed Angle Rotor for WX+ Ultraspeed Centrifuges
Product Code. 15315252
Change view
Click to view available options
Capacity:
8 x 100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
15315252 8 x 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15315252 Supplier Thermo Scientific™ Supplier No. 096087056
 Restricted Item
Spedizione stimata: 30-11-2026
Reduce the required number of runs for high-volume separations with this ultraspeed carbon fiber rotor that features two additional tube cavities.
Reduce the required number of runs for high-volume separations with the Thermo Scientific™ Fiberlite™ F37L-8 x 100 Fixed-Angle Rotor, an ultraspeed carbon fiber rotor that offers two additional tube cavities for an eight-place total capacity.
Fiberlite rotors:
  • Reduce run-times with higher G-forces achieved with lightweight carbon fiber rotors
  • Ensure structural integrity with corrosion- and fatigue-resistant design
  • Backed by a 15-year warranty
Ordering information:
Tube adapters are sold separately accommodating a variety of tubes in different capacities. Adapters available in packages of two. Contact your sales representative for more information.
Compatible with:
Thermo Scientific Sorvall™ WX Ultracentrifuge Series and Beckman™ Floor Model L Series (except Optima) centrifuges, Nalgene™ bottles and tubes
Recommended for:
Separations of subcellular organelles or concentration of viruses.
TRUSTED_SUSTAINABILITY
Specifications
Description Fiberlite F37L-8 x 100 Fixed-Angle Rotor
Material Carbon Fiber Composite
Type Fixed Angle Rotor
Capacity 8 x 100 mL
Max. RCF 182,460 x g
Max. Speed 37,000 rpm
Radius (Metric) 119 mm
Radius (Metric) Min. 48.8 mm
K Factor 168
Max. Tube Length 102 mm
Max. Tube Diameter 38 mm
Net Weight 25 lb. (11.34 kg)
For Use With (Equipment) Thermo Scientific™ Sorvall™ WX ultraCentrifuges
Angle 25°
Min. RCF 73,600 x g