Learn More
Eppendorf™ Filter Set for BioSpectrometer Fluorescence
Marca: Eppendorf™ 6137928009
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
Order Info
ForUseWithEquipment
Specifica
Filter Set | |
For verifying photometric accuracy and wavelength systematic error (according to NIST) and verifying fluorimetric precision (random error) and linearity | |
For BioSpectrometer fluorescence |