Learn More
Greiner Bio-One Filter Tips for Eppendorf
Prevents contamination with liquids and aerosols during pipetting process.
Marca: Greiner Bio-One 750288
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Non-cytotoxic
- Prevents aspiration of particles into interior of pipette
- Minimizes danger of unwanted cross-contamination
- Works with DNA and RNA and ra
Specifica
1000 μL | |
Sterile | |
Irradiation | |
No |
Filtered | |
Translucent | |
Anthos™ New Line, Costar™ Onepette™, Eppendorf™ Research™, Thermo Scientific™ Finnpipette™ Digital, Gilson™ Pipetman™, Gilson™ Pipetman™ P200, Human™ Pipettes | |
Polypropylene |