Learn More
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Finasteride is an inhibitor of the enzyme 5α-reductase, which converts testosterone to dihydrotestosterone (DHT)
- Finasteride also has been explored in neuroscience, as 5α-reductase enzymes are present in the central nervous system
- Finasteride has been seen to cross the blood-brain barrier and block production of neurosteroids
Applications
- Finasteride is mainly used in studies of prostate hyperplasia and tumors in mice, and can be studied in vitro or in vivo for its effects on hyperplasia, proliferation, and antioxidant activity
- Its activity has also been explored in brain, liver and testis tissues
- In vivo injection of finasteride has been shown to decrease neurogenesis and influence neuronal plasticity in mice
Specifica
Specifica
| Materiale o nome chimico | Finasteride |
| Punto di fusione | 252.0°C to 254.0°C |
| Spettro a infrarossi | Authentic |
| Intervallo percentuale saggio | 97.5% min. (HPLC) |
| Quantità | 5 g |
| Informazioni di solubilità | Solubility in water: very slightly soluble. |
| Peso formulazione | 372.54 |
| Percent Purity | 98% |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.