Learn More
DWK Life Sciences Fiolax™ Borosilicate Test Tube, with beaded rim
Codice prodotto. 10599841 Sfoglia Tutto DWK Life Sciences Prodotti
Cambia vista
Click to view available options
Capacità (sistema metrico):
2 mL
4 mL
6 mL
6.5 mL
16 mL
20 mL
25 mL
35 mL
39 mL
60 mL
80 mL
110 mL
Dimensione della confezione:
pezzi 100
pezzi 50
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) Dimensione della confezione
10599841 80 mL pezzi 50
10058180 6 mL pezzi 100
10333751 35 mL pezzi 100
10682401 39 mL pezzi 100
10091863 4 mL pezzi 100
10313561 2 mL pezzi 100
10282001 25 mL pezzi 100
10078190 16 mL pezzi 100
10333801 60 mL pezzi 50
10777381 6.5 mL pezzi 100
10221961 20 mL pezzi 100
10757571 110 mL pezzi 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Codice prodotto. 10599841 Fornitore DWK Life Sciences N. del fornitore 261103603
In stock
Test tubes are universal cylindrical laboratory glassware with a round bottom that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
  • Thin-walled and therefore resistant to rapid temperature changes or localized heating
  • Easier pouring due to beaded rim
  • Fired-on, highly durable white ceramic
Specifica
Descrizione 0.6 to 0.7 mm thickness
Capacità (sistema metrico) 80 mL
Materiale Fiolax
Da utilizzare con Heating of small amounts; Pretreatments; Storage of small amounts of liquid
Quantità 50/Pk.
Autoclavabile Autoclavable
Diametro (sistema metrico) 25 mm