missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ OptiGrow™ LB Broth Lennox Microbiological Media
Classic media in ready-to-use liquid form
Marca: Fisher BioReagents™ BP1431-10B1
Dettagli aggiuntivi : Peso : 10.00000kg
Descrizione
- Sterilized by autoclaving
- For high yield of bacterial culture
- No dusty powders to cause environmental concerns
- Fully automatic manufacturing process eliminates unexpected contamination
- LB Broth. Standard bacteriological nutrient medium for the propagation of E. coli for purposes of strain maintenance (sub-culturing), cloning, plasmid propagation, and protein expression
- Super Broth. Specifically developed to increase the yield of high copy number plasmids derived from suspension cultures of E. coli strains
- Terrific Broth. Supports higher cell densities than LB medium, increases plasmid DNA yield
Order Info
Supplied in case of ten 1L bottles
Specifica
Yellow | |
United States | |
10 x 1000 mL | |
10 x 1 L |
Liquid | |
LB Broth, Lennox | |
Bottle |