Learn More
Fisherbrand™ accumet™ Conductivity Calibration Kit
Product Code. 11580224 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
11580224 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11580224
  • ONLINE EXCLUSIVE PRICE
    336.00€
    243.97€ / pezzi 1
    Risparmia 92.03€
    27% Sconto
Spedizione stimata: 05-01-2027
Includes
  • Six precision resistors (nominal values 10ohm, 100ohm, 1kohm, 10kohm, 100kohm, and 1Mohm)
  • Connectors for accumet XL and AB conductivity meters
  • Foam padded case
Precision calibration resistor kit for accumet conductivity meters.
  • Supplied with a factory test certificate
TRUSTED_SUSTAINABILITY
Specifications
Type Conductivity Calibration Kit