Learn More
Fisherbrand™ Traceable™ Dual-Display Light Meter
Codice prodotto. 11784266 Sfoglia Tutto Fisherbrand Traceable Prodotti
Cambia vista
Click to view available options
Display:
LCD
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Display Dimensione della confezione
11784266 LCD Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11784266
  • ONLINE EXCLUSIVE PRICE
    307.00€
    244.30€ / pezzi 1
    Risparmia 62.70€
    20% Sconto
Visualizza tutte le promozioni
 Articolo soggetto a restrizioni
Spedizione stimata: 24-12-2026
Aggiungi al carrello
Aggiungi al carrello
Large LCD shows separate readings for lux and foot-candles in 1.375 in. H digits, plus selected source (daylight, tungsten, fluorescent or mercury).
Three Ranges
  • Measures lux from 0 to 1999, 2000 to 19,990 and 20,000 to 50,000 (with resolutions ranging from 1 to 100 lux)
  • Measures foot-candles from 0 to 199.9, 200 to 1999 and 2000 to 5000 (with resolutions ranging from 0.1 to 10 ft.-c)
  • Accuracy is ±4.5% of full scale reading
Features
  • Zero reference points for daylight, tungsten, fluorescent and mercury; can report percent differential from reference point
  • Photodiode and color-correction filter provide cosine- and color-corrected measurements
  • Push-button access to highest, lowest and average readings
  • HOLD function freezes display to capture readings
  • Optional Data Acquisition System (11-661-22) or Data Logger System (06-662-72) required to transfer readings to computer
Order Info
Dimensions: 7L x 3W x 1.5 in.H (17.8 x 7.6 x 3.8cm); Net weight: 9.5 oz. (269g); Easy-Use™ 115V adapter (11-661-23), Data Acquisition System (11-661-22) and Data Logger System (06-662-72) available as accessories
Certifications
With individually serial-numbered ISO 17025 certificate indicating instrument traceability to standards provided by NIST™.
Specifica
Tipo Light Meter
Precisione (footcandle) ±4.5% of full scale reading
Precisione (lux) ±4.5% of full scale reading
Certificazione/Conformità With individually serial-numbered ISO 17025 certificate indicating instrument traceability to standards provided by NIST™.
Altezza (sistema metrico) 3.2 cm
Include 2 in. Diameter (5.1cm) photocell, 40 in. (1m) cable, 9V alkaline battery, RS-232-C serial output for connection to IBM™-compatible computer or datalogger.
Lunghezza (sistema imperiale) 7 in.
Lunghezza (sistema metrico) 17.78 cm
Modello Dual Display
Intervallo (footcandle) 0 to 199.9, 200 to 1999 and 2000 to 5000
Intervallo (lux) 0 to 1999, 2000 to 19,990 and 20,000 to 50,000
Risoluzione (footcandle) 0.1 to 10 ft.-c
Risoluzione (lux) 1 to 100 lux
Larghezza (sistema imperiale) 3 in.
Larghezza (sistema metrico) 7.62 cm
Display LCD