Learn More
Fisherbrand™ Traceable™ Dual-Range Light Meter
Product Code. 11774266 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Display:
LCD
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Display Unit Size
11774266 LCD Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11774266
  • ONLINE EXCLUSIVE PRICE
    167.00€
    151.81€ / pezzi 1
    Risparmia 15.19€
    9% Sconto
View all promotions
 Restricted Item
Spedizione stimata: 04-01-2027
Add to basket
Add to basket
Measures both lux and foot-candles at a rapid sampling rate of 0.4 seconds. Display update rate is adjustable to 1 or 2 seconds.
  • Lux measurements: three ranges from 0 to 1999, 2000 to 19,990 and 20,000 to 50,000 lux; resolution from 1 to 100 lux; accuracy of ±5% plus two digits
  • Foot-candle measurements: three ranges from 0 to 199.9; 200 to 1999 and 2000 to 5000 ft-c.; resolution from 0.1 to 10 ft-c; accuracy of ±5% plus two digits
  • Push-button access to highest, lowest and average of recorded readings
  • DATA HOLD function 'freezes' display while analog output allows readings to be captured by a recorder
  • Photodiode and color-correction filter provide cosine- and color-corrected measurements
Order Info
Dimensions: 6.5L x 3W x 1.5 in. H (16.5 x 7.6 x 3.8cm). Net weight: 10 oz. (283g)
Certifications
Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Specifications
Type Light Meter
Accuracy (Foot Candle) ±5% plus two digits
Accuracy (Lux) ±5% plus two digits
Certifications/Compliance Individually serial-numbered Traceable Certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA™. It indicates traceability to standards provided by NIST™.
Height (Metric) 3.8 cm
Includes 2 in. Diameter (5.1cm) photocell, 40 in. (1m) cable, 9V alkaline battery and recorder output
Length (English) 6.5 in.
Length (Metric) 16.5 cm
Model Dual Range
Range (Foot Candle) 0 to 199.9; 200 to 1999 and 2000 to 5000 ft-c
Range (Lux) 0 to 1999, 2000 to 19,990 and 20,000 to 50,000 lux
Resolution (Foot Candle) 0.1 to 10 ft-c
Resolution (Lux) 1 to 100 lux
Width (English) 3 in.
Width (Metric) 7.62 cm
Display LCD